Skip to Content
Merck
All Photos(1)

Documents

233129

Sigma-Aldrich

2-(Trifluoromethyl)benzophenone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4COC6H5
CAS Number:
Molecular Weight:
250.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

59-62 °C (lit.)

SMILES string

FC(F)(F)c1ccccc1C(=O)c2ccccc2

InChI

1S/C14H9F3O/c15-14(16,17)12-9-5-4-8-11(12)13(18)10-6-2-1-3-7-10/h1-9H

InChI key

JXIWJBWMQXDALU-UHFFFAOYSA-N

General description

Assymmetric reduction of 2-(trifluoromethyl)benzophenone by lithium aluminium hydride treated with (S)-(+) or (R)-(−)-2 (2-iso-indolinyl)butan-1-ol has been investigated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparative synthesis of optically pure ortho-substituted benzhydrols by asymmetric reductions of the corresponding benzophenones.
Brown E, et al.
Tetrahedron Asymmetry, 3(7), 841-844 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service