226343
Ethyl bis(ethylthio)acetate
99%
Synonym(s):
Ethyl di(ethylthio)acetate, Glyoxylic acid ethyl ester diethyl mercaptal
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C2H5S)2CHCOOC2H5
CAS Number:
Molecular Weight:
208.34
Beilstein:
1767272
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
99%
refractive index
n20/D 1.497 (lit.)
bp
70 °C/0.1 mmHg (lit.)
density
1.071 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)C(SCC)SCC
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service