Skip to Content
Merck
All Photos(1)

Documents

226300

Sigma-Aldrich

Ethyl 1,3-dithiolane-2-carboxylate

98%

Synonym(s):

Glyoxylic acid ethyl ester ethylene mercaptal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2S2
CAS Number:
Molecular Weight:
178.27
Beilstein:
118157
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

refractive index

n20/D 1.539 (lit.)

bp

85 °C/0.1 mmHg (lit.)

density

1.249 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C1SCCS1

InChI

1S/C6H10O2S2/c1-2-8-5(7)6-9-3-4-10-6/h6H,2-4H2,1H3

InChI key

OMCSHTHLIQOHDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service