Skip to Content
Merck
All Photos(1)

Key Documents

221619

Sigma-Aldrich

1-(3-Phenylpropylamino)guanidine hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N4 · HCl
CAS Number:
Molecular Weight:
228.72
EC Number:
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Assay

98%

SMILES string

Cl.NC(=N)NNCCCc1ccccc1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3-phenylpropylaminoguanidine.HCl (43-522): a specific carbohydrate inhibitor in rats.
R S Ho et al.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 163(2), 200-205 (1980-02-01)
R S Ho et al.
The Journal of pharmacy and pharmacology, 32(8), 554-557 (1980-08-01)
The glucose absorption inhibitors, Phenformin and 3-phenylpropylaminoguanide HCl (SaH 43-522) have different effects on hepatic gluconeogenesis. Unlike phenformin, 43--522 in vivo does not inhibit hepatic gluconeogenesis; inhibition occurred only when 43--522 was added directly to a liver perfusion system. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service