Skip to Content
Merck
All Photos(1)

Documents

209775

Methyl 2,2-dichloro-1-methylcyclopropanecarboxylate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8Cl2O2
CAS Number:
Molecular Weight:
183.03
Beilstein:
1938341
MDL number:

Assay

99%

density

1.269 g/mL at 20 °C (lit.)

SMILES string

COC(=O)C1(C)CC1(Cl)Cl

InChI

1S/C6H8Cl2O2/c1-5(4(9)10-2)3-6(5,7)8/h3H2,1-2H3

InChI key

JGCAZFUFORGMFB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service