Skip to Content
Merck
All Photos(1)

Key Documents

196967

Sigma-Aldrich

2-Amino-5-trifluoromethyl-1,3,4-thiadiazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H2F3N3S
CAS Number:
Molecular Weight:
169.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

225-227 °C (lit.)

solubility

methanol: soluble 0.25 g/10 mL, clear, colorless

functional group

fluoro

SMILES string

Nc1nnc(s1)C(F)(F)F

InChI

1S/C3H2F3N3S/c4-3(5,6)1-8-9-2(7)10-1/h(H2,7,9)

InChI key

LTEUXHSAYOSFGQ-UHFFFAOYSA-N

Application

2-Amino-5-trifluoromethyl-1,3,4-thiadiazole was used in the synthesis of:
  • series of 2-sulfonamido/trifluoromethyl-6-(4′-substituted aryl/heteroaryl)imidazo[2,1-b]-1,3,4-thiadiazole derivatives
  • 2-trifluoromethyl/sulfonamido-5,6-diarylsubstituted imidazo[2,1-b]-1,3,4-thiadiazole derivatives
  • bicyclic bridgehead itrogen heterocycles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bridgehead nitrogen heterocycles. II. Formation by reaction of. alpha.-amino nitrogen heterocyclic compounds with chlorothioformyl chloride.
Pilgram K and Skiles RD.
The Journal of Organic Chemistry, 38(8), 1575-1578 (1973)
Andanappa K Gadad et al.
Bioorganic & medicinal chemistry, 12(21), 5651-5659 (2004-10-07)
A series of 2-sulfonamido/trifluoromethyl-6-(4'-substituted aryl/heteroaryl)imidazo[2,1-b]-1,3,4-thiadiazole derivatives (II) have been synthesized by reaction of 2-amino-5-sulfonamido/trifluoromethyl-1,3,4-thiadiazoles and an appropriate alpha-haloaryl/heteroaryl ketones. Further 5-bromo (III), 5-thiocyanato (IV), 5-gaunylhydrazone (V) derivatives were synthesized in order to study the effect of these substituents on biological
Andanappa K Gadad et al.
Bioorganic & medicinal chemistry, 16(1), 276-283 (2007-10-17)
A series of 2-trifluoromethyl/sulfonamido-5,6-diarylsubstituted imidazo[2,1-b]-1,3,4-thiadiazole derivatives 15a-j have been synthesized by the reaction of 2-amino-5-trifluoromethyl/sulfonamido-1,3,4-thiadiazoles 14a-b and appropriately substituted alpha-bromo-1,2-(p-substituted)diaryl-1-ethanones 13a-h. Structures of these compounds were established by IR, (1)H NMR, (13)C NMR, Mass, and HRMS data. The selected compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service