Skip to Content
Merck
All Photos(1)

Key Documents

194808

3,3,5-Trimethylhexahydroazepine,mixture of isomers

85%

Synonym(s):

3,3,5-Trimethyl-1-azacycloheptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19N
CAS Number:
Molecular Weight:
141.25
PubChem Substance ID:

Assay

85%

InChI

1S/C9H19N/c1-8-4-5-10-7-9(2,3)6-8/h8,10H,4-7H2,1-3H3

InChI key

YBRAVTNTXVZNMD-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service