190608
6-Benzoyl-2-naphthol
≥98%
Synonym(s):
6-Hydroxy-2-naphthyl phenyl ketone
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
≥98%
mp
161-162 °C (lit.)
solubility
ethanol: soluble 50 mg/mL
SMILES string
Oc1ccc2cc(ccc2c1)C(=O)c3ccccc3
InChI
1S/C17H12O2/c18-16-9-8-13-10-15(7-6-14(13)11-16)17(19)12-4-2-1-3-5-12/h1-11,18H
InChI key
MJKZGSBXCWNUHV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
6-Benzoyl-2-naphthol was used in the synthesis of potassium 6-benzoyl-2-naphthyl sulfate, substrate for histochemical investigation of aryl sulfatase activity. It was also used in preparation of substrate for histochemical demonstration of acid phosphatase in the organs of albino rat.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Histochemical demonstration of aryl sulfatase.
Science (New York, N.Y.), 116(3020), 539-543 (1952-11-14)
The histochemical demonstration of acid phosphatase by a post-incubation coupling technique.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 3(6), 455-470 (1955-11-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service