Skip to Content
Merck
All Photos(2)

Key Documents

17856

Sigma-Aldrich

N-Fmoc-iminodiacetic acid

≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17NO6
CAS Number:
Molecular Weight:
355.34
Beilstein:
7050417
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26

Assay

≥99.0%

mp

213-217 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)CN(CC(O)=O)C(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C19H17NO6/c21-17(22)9-20(10-18(23)24)19(25)26-11-16-14-7-3-1-5-12(14)13-6-2-4-8-15(13)16/h1-8,16H,9-11H2,(H,21,22)(H,23,24)

InChI key

LNHILYINDTUUMZ-UHFFFAOYSA-N

Other Notes

Reagent to introduce a glycine residue with free N- and C-termini by acylation (the anhydride is formed with TFA)[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H O Kim et al.
Journal of medicinal chemistry, 37(20), 3373-3382 (1994-09-30)
1,3-Dialkylxanthine analogues containing carboxylic acid and other charged groups on 8-position substituents were synthesized. These derivatives were examined for affinity in radioligand binding assays at rat brain A3 adenosine receptors stably expressed in CHO cells using the new radioligand [125I]AB-MECA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service