Skip to Content
Merck
All Photos(1)

Documents

178462

Sigma-Aldrich

2,4-Dichlorophenylacetonitrile

98%

Synonym(s):

2,4-Dichlorobenzyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CH2CN
CAS Number:
Molecular Weight:
186.04
Beilstein:
2047688
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

58-61 °C (lit.)

SMILES string

Clc1ccc(CC#N)c(Cl)c1

InChI

1S/C8H5Cl2N/c9-7-2-1-6(3-4-11)8(10)5-7/h1-2,5H,3H2

InChI key

VJARIBGMDPJLCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Dichlorophenylacetonitrile was used in the synthesis of 2,4-dichloromandelic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

348.8 °F - closed cup

Flash Point(C)

176 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Some α-alkoxyarylacetic acids.
Reeve W and Pickert PE.
Journal of the American Chemical Society, 79(8), 1932-1934 (1957)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service