Skip to Content
Merck
All Photos(1)

Key Documents

177644

Sigma-Aldrich

Thallium(I) bromide

99.999% trace metals basis

Synonym(s):

Thallium monobromide, Thallous bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TlBr
CAS Number:
Molecular Weight:
284.29
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

Assay

99.999% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: thallium

impurities

≤15.0 ppm Trace Metal Analysis

mp

>300 °C (lit.)

density

7.5 g/mL at 25 °C (lit.)

SMILES string

Br[Tl]

InChI

1S/BrH.Tl/h1H;/q;+1/p-1

InChI key

PGAPATLGJSQQBU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service