Skip to Content
Merck
All Photos(1)

Key Documents

16480

Sigma-Aldrich

4-Bromobiphenyl

technical, ≥90% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4Br
CAS Number:
Molecular Weight:
233.10
Beilstein:
1907453
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

technical

Assay

≥90% (GC)

form

solid

bp

310 °C (lit.)

mp

82-86 °C (lit.)
82-86 °C

solubility

dioxane: soluble 1 g/10 mL, clear, colorless to very faintly yellow

SMILES string

Brc1ccc(cc1)-c2ccccc2

InChI

1S/C12H9Br/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9H

InChI key

PKJBWOWQJHHAHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

In vitro metabolism of 4-bromobiphenyl by cytochrome P-450-dependent monooxygenases in rat hepatic microsomes has been investigated[1]. 4-Bromobiphenyl undergoes reduction to biphenyl in cationic micelles (cetyltrimethyl-ammonium bromide) by electrochemically generated anion radicals of 9-phenylanthracene[2].
may contain biphenyl and 4,4′-dibromobiphenyl

Application

4-Bromobiphenyl was used in the synthesis of 4-biphenylthiolate[3].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - Not applicable

Flash Point(C)

100 °C - Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrocatalytic reactions in organized assemblies: Part I. Reduction of 4-bromobiphenyl in cationic and non-ionic micelles.
Rusling JF, et al.
J. Electroanal. Chem. Interfac. Electrochem., 240(1), 201-216 (1988)
Structure of 4-biphenylthiolate on Au nanoparticle surfaces studied by UV-Vis absorption spectroscopy, transmission electron microscopy and surface-enhanced Raman scattering.
Jang S, et al.
Surface and Interface Analysis : SIA, 36(1), 43-48 (2004)
A D Davison et al.
Canadian journal of microbiology, 42(1), 66-71 (1996-01-01)
Sphingomonas paucimobilis BPSI-3 was previously isolated from a mixed microbial consortium growing on biphenyl as the sole source of carbon and energy. Transformation of 4-chlorobiphenyl (4CBP) was demonstrated by this strain, although little or no growth was observed. In minimal
A Parkinson et al.
Biochemical pharmacology, 31(10), 1849-1856 (1982-05-15)
The in vitro metabolism of biphenyl, 4-fluoro-, 4-chloro-, 4-bromo- and 4-iodobiphenyl by cytochrome P-450-dependent monooxygenases was investigated using hepatic microsomes from immature male rats pretreated with phenobarbitone or 3-methylcholanthrene. The major route of metabolism of biphenyl and the 4-halobiphenyls was
Man Wang et al.
Journal of chromatography. A, 1412, 12-21 (2015-08-19)
A new magnetic porous carbon-doped graphitic carbon nitride nanocomposite and experimental strategies were environment-friendly designed for solid phase extraction of brominated flame retardants from water sample. The easily synthesized and low cost nanocomposite was characterized using techniques, including Fourier transform

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service