All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H9N3S2
CAS Number:
Molecular Weight:
235.33
EC Number:
MDL number:
UNSPSC Code:
12352200
Assay
≥97%
mp
183-185 °C (dec.) (lit.)
SMILES string
S=C(Nc1ccccc1)Nc2nccs2
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Dopamine β-hydroxylase inhibitor.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service