Skip to Content
Merck
All Photos(1)

Documents

15495

Sigma-Aldrich

Boc-Sar-OH

≥99.0% (T)

Synonym(s):

Boc-N-methylglycine, Boc-sarcosine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCON(CH3)CH2COOH
CAS Number:
Molecular Weight:
189.21
Beilstein:
2046827
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:

Assay

≥99.0% (T)

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

88-90 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CN(CC(O)=O)C(=O)OC(C)(C)C

InChI

1S/C8H15NO4/c1-8(2,3)13-7(12)9(4)5-6(10)11/h5H2,1-4H3,(H,10,11)

InChI key

YRXIMPFOTQVOHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jing Zhang et al.
Cell death and differentiation, 27(5), 1569-1587 (2019-10-28)
Microtubule-targeting agents (MTAs) are a class of most widely used chemotherapeutics and their mechanism of action has long been assumed to be mitotic arrest of rapidly dividing tumor cells. In contrast to such notion, here we show-in many cancer cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service