Skip to Content
Merck
All Photos(1)

Key Documents

152935

Sigma-Aldrich

1-Piperidinepropanol

97%

Synonym(s):

1-(3-Hydroxypropyl)piperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17NO
CAS Number:
Molecular Weight:
143.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.4775 (lit.)

bp

94-95 °C/0.5 mmHg (lit.)

density

0.944 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OCCCN1CCCCC1

InChI

1S/C8H17NO/c10-8-4-7-9-5-2-1-3-6-9/h10H,1-8H2

InChI key

PLRXAFVBCHEMGD-UHFFFAOYSA-N

Application

1-Piperidinepropanol was used to prepare isothiourea[1].
Reactant in:
Mitsunobu reaction of intramolecular nitrile oxide-alkene cycloadditions
Suzuki-coupling

Reactant for synthesis of:
Histamine H3 receptor antagonists
Protein lysine methyltransferase G9a inhibitors
Serotonin-4 receptors antagonists
Checkpoint kinase Chk2 inhibitors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

208.0 °F - closed cup

Flash Point(C)

97.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shinya Harusawa et al.
Bioorganic & medicinal chemistry letters, 23(23), 6415-6420 (2013-10-22)
S-Alkyl-N-alkylisothiourea compounds containing various cyclic amines were synthesized in the search for novel nonimidazole histamine H3 receptor (H3R) antagonists. Among them, four N-alkyl S-[3-(piperidin-1-yl)propyl]isothioureas 18, 19, 22, and 23 were found to exhibit potent and selective H3R antagonistic activities against
Joseph A McQuail et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(50), 12537-12548 (2016-11-04)
Working memory, the ability to temporarily maintain representational knowledge, is a foundational cognitive process that can become compromised in aging and neuropsychiatric disease. NMDA receptor (NMDAR) activation in prefrontal cortex (PFC) is necessary for the pyramidal neuron activity believed to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service