Skip to Content
Merck
All Photos(1)

Key Documents

13145

Sigma-Aldrich

Benzyl-2-acetamido-4,6-O-benzylidene-2-deoxy-α-D-glucopyranoside

≥99.0% (sum of enantiomers, HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25NO6
CAS Number:
Molecular Weight:
399.44
Beilstein:
55951
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (sum of enantiomers, HPLC)

optical activity

[α]20/D +115±3°, c = 1% in pyridine

mp

256-261 °C (dec.)

SMILES string

CC(=O)N[C@@H]1[C@@H](O)[C@@H]2OC(OC[C@H]2O[C@@H]1OCc3ccccc3)c4ccccc4

InChI

1S/C22H25NO6/c1-14(24)23-18-19(25)20-17(13-27-21(29-20)16-10-6-3-7-11-16)28-22(18)26-12-15-8-4-2-5-9-15/h2-11,17-22,25H,12-13H2,1H3,(H,23,24)/t17-,18-,19-,20-,21?,22+/m1/s1

InChI key

NXGXFAKJUWEFEC-NVZUTRPHSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Carbohydrate building block; used for the synthesis of N-acetyl muramic acid and similar compounds; preparation of derivatives of 2,3-diamino-2,3-dideoxy-glucopyranoside.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Imoto et al.
Bulletin of the Chemical Society of Japan, 59, 3207-3207 (1986)
[Diamino sugars. X. Notes on a simple synthesis of 2,3-diamino-2,3-didesoxy-D-glucose].
W Meyer zu Reckendorf
Chemische Berichte, 102(12), 4207-4208 (1969-01-01)
AN IMPROVED, STEREOSELECTIVE SYNTHESIS OF 2-AMINO-3-O-(D-1-CARBOXYETHYL)-2-DEOXY-D-GLUCOSE (MURAMIC ACID).
T OSAWA et al.
The Journal of organic chemistry, 30, 448-450 (1965-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service