Skip to Content
Merck
All Photos(1)

Key Documents

126403

Sigma-Aldrich

2,5-Dimethylbenzyl chloride

98%

Synonym(s):

1-Chloromethyl-2,5-dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3CH2Cl
CAS Number:
Molecular Weight:
154.64
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.537 (lit.)

bp

221-226 °C (lit.)

density

1.046 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Cc1ccc(C)c(CCl)c1

InChI

1S/C9H11Cl/c1-7-3-4-8(2)9(5-7)6-10/h3-5H,6H2,1-2H3

InChI key

PECXPZGFZFGDRD-UHFFFAOYSA-N

General description

2,5-Dimethylbenzyl chloride undergoes low-temperature Friedel-Crafts step-growth polymerization reaction. It has been studied for the effect of reaction conditions on polymer molecular weight, linearity, glass transition temperature, and crystalline properties.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service