Merck
  • Bactericidal activity of carvacrol towards the food-borne pathogen Bacillus cereus.
Bactericidal activity of carvacrol towards the food-borne pathogen Bacillus cereus.
Journal of applied microbiology (1998-09-29)
A Ultee, L G Gorris, E J Smid
ABSTRACT
Carvacrol, a natural plant constituent occurring in oregano and thyme, was investigated for its bactericidal effect towards the food-borne pathogen Bacillus cereus. Carvacrol showed a dose-related growth inhibition of B. cereus. At concentration of 0.75 mmol l-1 and above, total inhibition of the growth was observed. Below this concentration, carvacrol extended the lag-phase, reduced the specific growth rate and reduced the maximum population density. Incubation for 40 min in the presence of 0.75-3 mmol l-1 carvacrol decreased the number of viable cells of B. cereus exponentially. Spores were found to be approximately 2-3 fold less sensitive to carvacrol than vegetative cells. Bacillus cereus cells showed reduced susceptibility towards carvacrol at pH 7.0 compared with different values between pH 4.5 and 8.5. The culture and exposure temperatures had a significant influence on the survival of vegetative cells. The highest death rate of cells was observed at an exposure temperature of 30 degrees C. Membrane fluidity was found to be an important factor influencing the bactericidal activity of carvacrol.
Sigma-Aldrich
Carvacrol, 98%