MilliporeSigma
  • Development and characterization of antimicrobial poly(l-lactic acid) containing trans-2-hexenal trapped in cyclodextrins.
Development and characterization of antimicrobial poly(l-lactic acid) containing trans-2-hexenal trapped in cyclodextrins.
International journal of food microbiology (2011-12-20)
Min Jung Joo, Crispin Merkel, Rafael Auras, Eva Almenar
ABSTRACT
Trans-2-hexenal, a naturally occurring plant volatile with antimicrobial capacity, was encapsulated into β-cyclodextrins (β-CDs), enzymatically modified starch, and shown effective to control main microorganisms causing food spoilage (Alternaria solani, Aspergillus niger, Botrytis cinerea, Colletotrichum acutatum, Penicillium sp). Loaded β-CDs were incorporated into a poly(L-lactic acid) (PLA) matrix by extrusion and casting, and yielded antimicrobial polymers made from natural resources. A masterbatch was used prior to sheet casting to improve the dispersion of the antimicrobial agent in the PLA matrix. However, this increased the number of extrusion processes for the material. The concentration of the antimicrobial compound in the polymers and its antimicrobial capacity against one food spoilage microorganism (A. solani) were measured during the different processing operations. Although the concentration of trans-2-hexenal was reduced by processing by about 70 and 99% compared to the loaded β-CDs, for the masterbatch and sheet, respectively, the polymers were still effective in reducing microbial growth. The changes of the polymer properties due to the addition of the antimicrobial agent were investigated, too. It was found that the mechanical and barrier properties of the PLA were changed (decreased by about half the tensile strength and elongation at break and nine-fold increased permeability) while the physical properties remained the same. Based on these results, the developed polymer may be a viable antimicrobial material for applications in food packaging.
Sigma-Aldrich
trans-2-Hexen-1-al, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
trans-2-Hexen-1-al, ≥95%, FCC, FG