Merck
Therapeutics targeting the fibrinolytic system.
Experimental & molecular medicine (2020-03-11)
Haili Lin, Luning Xu, Shujuan Yu, Wanjin Hong, Mingdong Huang, Peng Xu
ABSTRACT
The function of the fibrinolytic system was first identified to dissolve fibrin to maintain vascular patency. Connections between the fibrinolytic system and many other physiological and pathological processes have been well established. Dysregulation of the fibrinolytic system is closely associated with multiple pathological conditions, including thrombosis, inflammation, cancer progression, and neuropathies. Thus, molecules in the fibrinolytic system are potent therapeutic and diagnostic targets. This review summarizes the currently used agents targeting this system and the development of novel therapeutic strategies in experimental studies. Future directions for the development of modulators of the fibrinolytic system are also discussed.
Sigma-Aldrich
δ-Valerolactam, 98%