
Memantine Related Compound A

Synonym(s):
1,3-Dimethyladamantane, 1,3-Dimethyltricyclo[3.3.1.13,7]decane
Empirical Formula (Hill Notation):
C12H20
CAS Number:
Molecular Weight:
164.29
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2