
Betaine aldehyde chloride

Synonym(s):
(Formylmethyl)trimethylammonium chloride
Empirical Formula (Hill Notation):
C5H12ClNO
CAS Number:
Molecular Weight:
137.61
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1