Key Documents
Z742780
smartPans cahn style aluminim weighing dish
pack of 100 (Dishes supplied in flip top vial)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.35
material
aluminum
packaging
pack of 100 (Dishes supplied in flip top vial)
manufacturer/tradename
LevGo Inc
diam. × H × thickness
25 mm × 3.59 mm × 0.038 mm
weight
53 mgEach boat
General description
Ideal for microbalances, organic micro elementary / thermal analysis, and pharmaceutical use.
  • Made of Ultra High Vacuum (UVH) Foil - “A” wettable and completely free from oils normally found on other grades of aluminum foil.
  • Foil meets ASTM B-479, specification 3.1.4 and 10.3.1.
  • “Cahn style” accessory pan / disc format
  • Compatible with most commercially available analyzers and microbalances
  • Foil and Pans are made in the USA
  • Packaged 20 or 100 per easy-to-use PP flip top vials - Both the pans and the vials recyclable
Legal Information
smartPans is a trademark of LevGo Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)