Z327484
BRAND® graduated cylinder, PMP, embossed scale
volume 2000 mL, accuracy: 20 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Agency
ISO 6706 (meets DIN 12 680-1 tolerances for Class B cylinders)
feature
subdivision 20 mL
manufacturer/tradename
BRAND
volume measuring range
2000 mL, accuracy: 20 mL
H
535 mm
Looking for similar products? Visit Product Comparison Guide
General description
Graduated cylinders, tall form, with embossed scale, are made from transparent PMP. To DIN 12681 and ISO 6706. Calibrated to contain (TC, In). Perfect cylindrical design and equal graduation marks allow for easy volume reading. Functional spout minimizes spills. Exposure to temperatures up to 121 °C (autoclaving) will not cause permanent exceeding of error limits. Cleaning temperatures below 60 °C is recommended.
Other Notes
BR34964 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service