MTCP81005S
MTC™ Bio Reservoir
capacity 100 mL, sterile, pack of 200 ea (40 x bags 5 ea)
Synonym(s):
reagent dish, reagent holder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121522
NACRES:
NB.12
Recommended Products
sterility
sterile
packaging
pack of 200 ea (40 x bags 5 ea)
manufacturer/tradename
MTC™ Bio P8100-5S
capacity
100 mL
Looking for similar products? Visit Product Comparison Guide
General description
Reservoir, 100ml sterile, 5/sterile bag, 200/cs
Features and Benefits
- Graduated inner walls
- Micro-trough U-shaped bottoms
- Pouring spouts on all 4 corners
Legal Information
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service