MT30671545
Mettler Toledo SevenDirect SD50 pH / Ion Meter F-Ion Kit
electrode, perfectION Fluoride
Synonym(s):
Benchtop pH meter, fluoride Ion meter, digital pH meter
About This Item
AC/DC input
, universal plug set
220 V / 240 V
feature
electrode perfectION Fluoride
packaging
pack of 1 kit
manufacturer/tradename
Mettler-Toledo 30671545
concentration measuring range
-2.000-20.000 pX, resolution: 3 or 4 digits (ion)
0-999,999 mg/L (ion)
mV measuring range
-2000-2000 mV, resolution: 1; 0.1
pH measuring range
accuracy: ±0.002, resolution: 0.1
resolution: 0.001
resolution: 0.01
temperature measuring range
-30-130 °C, resolution: 0.1 °C
dilution
(pH and Ion measurement)
General description
Large color touchscreen provides direct measurement and easy data entry. Instructions guide users through the sensor calibration process.
Tracks your Records
The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
Tough Enough
Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
Combined Fluoride sensor
The perfectION™ F sensor is a combined fluoride ion sensor with an integrated reference electrode. The special Click & Clear™ junction allows for an optimal contact of the electrolyte solution and the sample, making measurement and maintenance easy.
Electrode Arm for Efficient Operation
The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy. Dedicated positions for cleaning and storage support correct sensor handling. Vertical and horizontal movements facilitate precise positioning, even with one hand.
Protect Critical Settings
A two-level user management protects the most important settings. Operators retain the flexibility to individually set preferences like language and screen brightness.
Display Adapts to Your Needs
View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
Connectors - BNC; RCA / Cinch; 2 mm Reference; USB A (x2); USB B; Mini DIN; (for stirrer)
Application
Features and Benefits
- Large color touchscreen provides direct measurement and easy data entry.
- The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
- Smart design and cover keep the instrument safe from dust and spills
- Click & Clear™ junction allows for an optimal contact of the electrolyte solution and the sample, making measurement and maintenance easy.
- Electrode arm for efficient operation
- Protect critical settings Protected with two-level user management
- Display adapts to your needs to maximize readability
Components
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service