Skip to Content
Merck
All Photos(3)

Key Documents

HPA022978

Sigma-Aldrich

Anti-KRT24 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CK-24, Anti-Cytokeratin-24, Anti-K24, Anti-Keratin, type I cytoskeletal 24, Anti-Keratin-24, Anti-Type I keratin-24

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

LDIKTRLEVEIETYRRLLDGEGGGSSFAEFGGRNSGSVNMGSRDLVSGDSRSGSCSGQGRDSSKTRVTKTIVEELVDGKVVSSQVSSIS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT24(192666)

Immunogen

Keratin, type I cytoskeletal 24 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Keratins are structural proteins that are crucial for the formation of keratin intermediate filaments. KRT24 is a type 1 epithelial keratin. It is expressed in keratinocytes, placenta, colon and spleen. It is associated with the Naegeli-Franceschetti-Jadassohn syndrome, autosomal dominant ectodermal dysplasia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75783

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Naeem et al.
The British journal of dermatology, 155(6), 1184-1190 (2006-11-17)
Ectodermal dysplasias (EDs) describe a large and complex group of disorders characterized by abnormal development of the skin and appendages (hair, nails, teeth and sweat glands). Of the approximately 200 different EDs, about 30 have been studied at the molecular
B Jerábková et al.
The British journal of dermatology, 162(5), 1004-1013 (2009-12-25)
Epidermolysis bullosa simplex (EBS) is an inherited skin disorder caused by mutations in the keratin 5 (KRT5) and keratin 14 (KRT14) genes, with fragility of basal keratinocytes leading to epidermal cytolysis and blistering. In this study, we characterized mutations in
Eli Sprecher et al.
The Journal of investigative dermatology, 119(3), 692-698 (2002-09-17)
Naegeli-Franceschetti-Jadassohn syndrome and dermatopathia pigmentosa reticularis are autosomal dominant ectodermal dysplasias characterized by the absence of dermatoglyphics, reticulate hyper pigmentation of the skin, hypohidrosis, and heat intolerance. Palmoplantar keratoderma, nail dystrophy, and enamel defects are common in Naegeli-Franceschetti-Jadassohn syndrome, whereas
Kiran Kc et al.
PloS one, 10(6), e0127755-e0127755 (2015-06-04)
Epithelial differentiation is an essential physiological process that imparts mechanical strength and barrier function to squamous epithelia. Perturbation of this process can give rise to numerous human diseases, such as atopic dermatitis, in which antigenic stimuli can penetrate the weakened

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service