EP0030601572
Eppendorf® Microplate 96/U-PP, 96 wells
gray polypropylene, round bottom white wells, PCR clean, pkg of 80 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122107
NACRES:
NB.15
material:
gray polypropylene
round bottom white wells
round bottom white wells
size:
96 wells
sterility:
sterile; autoclaved
Recommended Products
material
gray polypropylene
round bottom white wells
sterility
sterile; autoclaved
packaging
pkg of 80 ea
manufacturer/tradename
Eppendorf® 0030601572
technique(s)
PCR: suitable
size
96 wells
Looking for similar products? Visit Product Comparison Guide
General description
Pipette, check and recover your samples easily with Eppendorf round bottom Microplates. The easy-to-read OptiTrack alphanumeric matrix label for identifying each well speeds up filling and significantly reduces the risk of pipetting errors.
Application
- sample storage and preparationassays that require high resistance against temperature or solvents
- active ingredient screening
- combinatorial chemistry
- storage of cDNA or genomic banks
- protein analysis
- genotyping
- storage of siRNA or oligonucleotide libraries
- preparation of tissue and plant lysates
Features and Benefits
- Eppendorf Microplates consist of polypropylene wells which enhance sample visibility. In addition, polypropylene also ensures resistance towards chemicals, mechanical stressors and high temperatures.
- A raised well rim and a smooth surface facilitates secure sealing.
- Eppendorf Microplates can withstand centrifugal forces up to six thousand times the gravitational force.
- The OptiTrack matrix feature enables faster well identification and reduces pipetting error.
- An optimized RecoverMax well geometry minimizes the remaining volume and ensures smooth mixing properties.
- Eppendorf Microplates function efficiently over a wide temperature range, ranging from -86 °C to 100 °C.
- These microplates can be autoclaved at 121 °C for 20 min.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service