Skip to Content
Merck
All Photos(1)

Key Documents

CLS3700

Corning® 384 well microplate

clear polystyrene, flat bottom, non-sterile, lid: no, case of 100

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
clear bottom
clear polystyrene
flat bottom
well volume:
112 μL
well working volume:
20-80 μL
wells:
384 wells
Pricing and availability is not currently available.

material

clear bottom
clear polystyrene
flat bottom

sterility

non-sterile

feature

lid: no
skirt

packaging

case of 100

manufacturer/tradename

Corning 3700

well volume

112 μL

well working volume

20-80 μL

wells

384 wells

color

clear

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 384 well microplate is ideal for immunoassays
  • High binding surface binds medium (>10kDa) and large biomolecules with ionic groups and/or hydrophobic regions
  • ELISA microplates with binding capacity is approximately 400 to 500ng IgG/cm²
  • Standard microplate footprint and dimensions

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service