Skip to Content
Merck
All Photos(1)

Key Documents

CL11400

Sigma-Aldrich

DMT-locMeC(bz) Phosphoramidite

Synonym(s):

DMT-locMeC(bz) Phosphoramidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H54N5O9P
CAS Number:
Molecular Weight:
875.94
UNSPSC Code:
41116105

biological source

non-animal source (BSE/TSE no Risk)

Quality Level

product line

Proligo Reagents

Assay

≥98.0% (31P-NMR)
≥98.0% (reversed phase HPLC)

form

powder

impurities

≤3 wt. % Residual Solvent Content
<0.4 wt. % water content (Karl Fischer)
<0.5% single unspecified Impurity (reversed phase HPLC)

color

white to off-white

solubility

acetonitrile: 0.2 M, clear, colorless

absorption

<0.1 in acetonitrile at 0.2 m

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service