Skip to Content
Merck
All Photos(1)

Documents

CRM361

Supelco

TPH - Sea Sediment 1

pkg of 100 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

feature

Certified TPH Concentration - 695 mg/Kg

packaging

pkg of 100 g

manufacturer/tradename

RTC CRM361-100

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

matrix

soil

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service