Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
87576
Color Reference Solutions acc. to USP
analytical standard, set of 20 × 2 mL (20 standard solution)
About This Item
Recommended Products
grade
analytical standard
Quality Level
Agency
according to USP
shelf life
limited shelf life, expiry date on the label
packaging
set of 20 × 2 mL (20 standard solution)
application(s)
cleaning products
coatings
cosmetics
environmental
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical
format
multi-component solution
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Ready-to-use solutions
- Prepared and qualified in accordance with United States Pharmacopoeia (USP)
- Available in tamper-evident 2 mL ampules
- Expiry dated on the label with a limited shelf life
- Accompanied with a detailed certificate of analysis for an effective quality management system
Components
- Colour Reference Solution A - 2mL ampule
- Colour Reference Solution B - 2mL ampule
- Colour Reference Solution C - 2mL ampule
- Colour Reference Solution D - 2mL ampule
- Colour Reference Solution E - 2mL ampule
- Colour Reference Solution F - 2mL ampule
- Colour Reference Solution G - 2mL ampule
- Colour Reference Solution H - 2mL ampule
- Colour Reference Solution I - 2mL ampule
- Colour Reference Solution J - 2mL ampule
- Colour Reference Solution K - 2mL ampule
- Colour Reference Solution L - 2mL ampule
- Colour Reference Solution M - 2mL ampule
- Colour Reference Solution N - 2mL ampule
- Colour Reference Solution O - 2mL ampule
- Colour Reference Solution P - 2mL ampule
- Colour Reference Solution Q - 2mL ampule
- Colour Reference Solution R - 2mL ampule
- Colour Reference Solution S - 2mL ampule
- Colour Reference Solution T - 2mL ampule
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Met. Corr. 1
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service