Skip to Content
Merck
All Photos(1)

Key Documents

74099

Supelco

Tryptamine hydrochloride

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2-(3-Indolyl)ethylamine hydrochloride, 3-(2-Aminoethyl)indole hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2 · HCl
CAS Number:
Molecular Weight:
196.68
Beilstein:
3568419
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

crystals

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

mp

253-255 °C (lit.)

SMILES string

Cl[H].NCCc1c[nH]c2ccccc12

InChI

1S/C10H12N2.ClH/c11-6-5-8-7-12-10-4-2-1-3-9(8)10;/h1-4,7,12H,5-6,11H2;1H

InChI key

KDFBGNBTTMPNIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 . This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Reactant for preparation of:
  • Tryptamine derivatives as inhibitors against hepatitis B virus
  • Carbamoyl epipodophyllotoxins as potential antitumor agents
  • Anthranilic acid derivatives as CCK receptor antagonists
  • Brassinin derivatives as indoleamine 2,3-dioxygenase inhibitors
  • Antispasmodic agents
  • β-carbolinium cations as new antimalarial agents

Biochem/physiol Actions

Vasoactive; may have a neuromodulator function.

Other Notes

Biogenic amine formed from the decarboxylation of tryptophan by L-aromatic amino acid decarboxylase.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the purity on this? Thanks. I don't understand the sample CoA. Is it 80.7% pure.

    1 answer
    1. The purity reported in the Certificate of Analysis is for the Tryptamine only. The sample document reports a lot purity of 80.7% tryptamine. Please navigate to the ‘DOCUMENTATION’ section of the Product Detail Page to access a Certificate:
      https://www.sigmaaldrich.com/product/sial/74099#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service