XX42PFK01
Pellicon® Mini Holder Fitting Kit Polypropylene
All Photos(1)
About This Item
UNSPSC Code:
23151800
eCl@ss:
32031690
Recommended Products
General description
The Pellicon® Mini Holder Fitting Kit Polypropylene is an accessory to the Pellicon® Mini Cassette Holder which has a simple design which makes process development and small scale pharmaceutical manufacturing simple and efficient.
The Pellicon® Mini holder operates one to three Pellicon® 2 Mini cassettes in parallel or Pellicon® 3 cassettes. This sanitary holder is tightened with a small torque wrench to compress the cassettes between a manifold plate that conveys fluids in and out of the cassettes and an end plate that seals the cassettes together.
The Mini holder is designed for process development and small volume pharmaceutical manufacturing.
The Pellicon® Mini holder operates one to three Pellicon® 2 Mini cassettes in parallel or Pellicon® 3 cassettes. This sanitary holder is tightened with a small torque wrench to compress the cassettes between a manifold plate that conveys fluids in and out of the cassettes and an end plate that seals the cassettes together.
The Mini holder is designed for process development and small volume pharmaceutical manufacturing.
Features and Benefits
The Mini holder and Pellicon® Mini Holder Fitting Kit Polypropylene is designed for process development and small volume pharmaceutical manufacturing.
Legal Information
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany