Skip to Content
Merck
All Photos(1)

Key Documents

ABE1934

Sigma-Aldrich

Anti-Gcn5 Antibody

from rabbit

Synonym(s):

Histone acetyltransferase KAT2A, General control of amino acid synthesis protein 5-like 2, hGCN5, Histone acetyltransferase GCN5, HsGCN5, Lysine acetyltransferase 2A, STAF97

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

ChIP: suitable (ChIP-seq)
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KAT2A(2648)

General description

Histone acetyltransferase KAT2A (EC 2.3.1.48; UniProt Q92830; also known as General control of amino acid synthesis protein 5-like 2, hGCN5, Histone acetyltransferase GCN5, HsGCN5, Lysine acetyltransferase 2A, STAF97) is encoded by the KAT2A (also known as GCN5, GCN5L2, HGCN5) gene (Gene ID 2648) in human. Histone acetyltransferases (HATs) are transcription co-activators recruited by transcriptional activators to target gene promoters, where they enhance the transcription activity by acetylating lysine residues within the N-terminal regions of histones. KAT2A/KAT2B, MYST (MOZ, Ybf2 (Sas3), Sas2, and Tip60), TAFII250, CREBBP/EP300, and SRC represent the five HAT families. In addtion to functioning as transcription co-activators, KAT2A (GCN5) and KAT2B (PCAF) are also reported to repress IFN‐β production and innate antiviral immunity by inhibiting the innate immune signaling kinase TBK1 in the cytoplasm in a HAT‐independent and non‐transcriptional manner.

Specificity

This polyclonal antibody was raised against human KAT2A isoform GCN5-L N-terminal sequence that is 95% conserved in murine and rat Kat2a/Gcn5, but missing in human spliced isoform GCN5-S (UniProt Q92830-2).

Immunogen

Epitope: N-terminus.
Ovalbumin-conjugated linear peptide corresponding to the N-terminal sequence of human Gcn5.

Application

Anti-Gcn5 Antibody is an antibody against Gcn5 for use in Immunoprecipitation, Western Blotting, ChIP-seq, Chromatin Immunoprecipitation (ChIP).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins
Western Blotting Analysis: 1.2 µg/mL from a representative lot detected Gcn5 immunoprecipited from HeLa nuclear extract.
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot immunoprecipitated KAT2A/GCN5-associated DNA fragments by ChIP using chromatin preparations from murine CH12 and human GM12878 lymphoblasts (Cheng, Y., et al. (2014). Nature. 515(7527):371-375).
ChIP-seq Analysis: A representative lot was employed to generate Gcn5 genome-wide binding profiles by ChIP-seq analysis using chromatin preparations from murine CH12 and human GM12878 lymphoblasts (Cheng, Y., et al. (2014). Nature. 515(7527):371-375).

Quality

Evaluated by Immunoprecipitation in HeLa nuclear extract.

Immunoprecipitation Analysis: 10 µg of this antibody immunoprecipited Gcn5 from 500 µg of HeLa nuclear extract.

Target description

~100 kDa observed. 93.93 kDa (human GCN5-L; UniProt Q92830-1) calculated.

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service