Skip to Content
Merck
All Photos(1)

Key Documents

AB1673

Sigma-Aldrich

Anti-Leptin Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
clone:
polyclonal
application:
RIA
WB
species reactivity:
mouse
technique(s):
radioimmunoassay: suitable
western blot: suitable
citations:
2

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

radioimmunoassay: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Lep(16846)

Specificity

Reacts with mouse leptin.

Immunogen

Recombinant mouse leptin.

Application

Anti-Leptin Antibody detects level of Leptin & has been published & validated for use in RIA & WB.
Research Category
Metabolism
Research Sub Category
Lipid Metabolism & Weight Regulation
Western blot: 1:1,000

RIA: 1:1,000

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: AB3385P

Physical form

Format: Purified
Purified immunoglobulin in PBS. Contains no preservative.

Storage and Stability

Maintain at -20°C in diluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasser Majeed et al.
Scientific reports, 11(1), 8177-8177 (2021-04-16)
The NAD+-dependent deacetylase SIRT1 controls key metabolic functions by deacetylating target proteins and strategies that promote SIRT1 function such as SIRT1 overexpression or NAD+ boosters alleviate metabolic complications. We previously reported that SIRT1-depletion in 3T3-L1 preadipocytes led to C-Myc activation
Qiqi Yang et al.
Disease models & mechanisms, 12(2) (2019-02-06)
Cancer cachexia affects up to 80% of patients with advanced solid cancer and leads to excessive muscle wasting. Here, using an inducible zebrafish hepatocellular carcinoma (HCC) model driven by oncogenic krasG12V , we observed a progressive muscle-wasting phenotype in adult

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service