Skip to Content
Merck
All Photos(1)

Key Documents

8.01395

Sigma-Aldrich

2-Butoxyethyl acetate

for synthesis

Synonym(s):

2-Butoxyethyl acetate, Ethylene glycol monobutyl ether acetate, Ethylene glycol butyl ether acetate, Acetic acid 2-butoxyethyl ester, Butyl glycol acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2O(CH2)3CH3
CAS Number:
Molecular Weight:
160.21
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-933-3
NACRES:
NA.22

vapor pressure

0.31 hPa ( 20 °C)

Quality Level

Assay

≥98% (GC)

form

liquid

autoignition temp.

280 °C

potency

2400 mg/kg LD50, oral (Rat)
1500 mg/kg LD50, skin (Rabbit)

expl. lim.

1.7-8.4 % (v/v)

pH

4 (20 °C, 10 g/L in H2O)

bp

194-196 °C/1013 hPa

mp

-64--62 °C

transition temp

flash point 78 °C

density

0.94 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H16O3/c1-3-4-5-10-6-7-11-8(2)9/h3-7H2,1-2H3

InChI key

NQBXSWAWVZHKBZ-UHFFFAOYSA-N

Application



  • Three-Dimensional Printing of Poly(glycerol sebacate) Acrylate Scaffolds via Digital Light Processing.: This research demonstrates the use of 2-butoxyethyl acetate in the solvent processing part of 3D printing polymers, suggesting its potential in biomedical applications and materials science (Wu YL et al., 2020).



Analysis Note

Assay (GC, area%): ≥ 98 %
Density (d 20 °C/ 4 °C): 0.940 - 0.942
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service