Skip to Content
Merck
All Photos(1)

Key Documents

681525

Sigma-Aldrich

Wiskostatin

A cell-permeable N-alkylated carbazole derivative that selectively blocks actin filament assembly.

Synonym(s):

Wiskostatin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18Br2N2O
CAS Number:
Molecular Weight:
426.15
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Brc1cc2c([n](c3c2cc(cc3)Br)CC(O)CN(C)C)cc1

InChI

1S/C17H18Br2N2O/c1-20(2)9-13(22)10-21-16-5-3-11(18)7-14(16)15-8-12(19)4-6-17(15)21/h3-8,13,22H,9-10H2,1-2H3

InChI key

XUBJEDZHBUPBKL-UHFFFAOYSA-N

General description

A cell-permeable N-alkylated carbazole derivative that selectively blocks actin filament assembly. Acts as a selective, reversible inhibitor of N-WASP (neural Wiskott Aldrich syndrome protein), a signal integrating protein. Appears to bind to N-WASP, stabilize the autoinhibited conformation and prevent the activation of Arp2/3 (actin-related protein 2/3) complex.
A cell-permeable N-alkylated carbazole derivative that selectively blocks actin filament assembly. Acts as a selective, reversible inhibitor of neural Wiskott Aldrich Syndrome protein (N-WASP). Reported to bind to N-WASP, stabilize the auto-inhibited conformation, and prevent the activation of actin-related protein 2/3 (Arp2/3) complex.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
N-WASP (neural Wiskott Aldrich syndrome protein)
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Peterson, J.R., et al. 2004. Nat. Struct. Mol. Biol.11, 747.
Peterson, J.R., and Mitchison, T.J. 2002. Chem. Biol.9, 1275.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service