Skip to Content
Merck
All Photos(1)

Key Documents

454180

Sigma-Aldrich

2-Methoxyestradiol

A natural metabolite of 17β-estradiol that is devoid of estrogenic activity.

Synonym(s):

2-Methoxyestradiol, 2-Methoxy-3,17β-dihydroxyestra-1,3,5(10)-triene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O3
CAS Number:
Molecular Weight:
302.41
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥90% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: soluble
ethanol: soluble

shipped in

ambient

storage temp.

10-30°C

SMILES string

O[C@@H]1[C@@]2([C@H]([C@H]3[C@H](CC2)c4c(cc(c(c4)OC)O)CC3)CC1)C

InChI

1S/C19H26O3/c1-19-8-7-12-13(15(19)5-6-18(19)21)4-3-11-9-16(20)17(22-2)10-14(11)12/h9-10,12-13,15,18,20-21H,3-8H2,1-2H3/t12-,13+,15-,18-,19-/m0/s1

InChI key

CQOQDQWUFQDJMK-SSTWWWIQSA-N

General description

A natural metabolite of 17β-estradiol that is devoid of estrogenic activity. Inhibits cell proliferation and angiogenesis. Markedly inhibits neovascularization of solid tumors and suppresses tumor growth in mice. Binds to the colchicine binding site of tubulin and has been suggested to function as a natural regulator of microtubule assembly and function.
A natural metabolite of 17β-estradiol that is devoid of estrogenic activity. Inhibits cell proliferation and angiogenesis. Markedly inhibits the neovascularization of solid tumors and suppresses tumor growth in mice. Binds to the colchicine binding site of tubulin, and has been suggested to function as a natural regulator of microtubule assembly and function.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Inhibits cell proliferation and angiogenesis
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Toxic (F)

Other Notes

Aizu-Yokata, E., et al. 1995. Cancer Res.55, 1863.
Cushman, M., et al. 1995. J. Med. Chem. 38, 2041.
D’Amato, R.J., et al. 1994. Proc. Natl. Acad. Sci. USA 91, 3964.
Forsis, T., et al. 1994. Nature368, 237.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service