Skip to Content
Merck
All Photos(1)

Key Documents

324832

Sigma-Aldrich

EGFR Inhibitor II, BIBX1382

The EGFR Inhibitor II, BIBX1382, also referenced under CAS 196612-93-8, controls the biological activity of EGFR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

EGFR Inhibitor II, BIBX1382, N⁸-(3-Chloro-4-fluorophenyl)-N²-(1-methylpiperidin-4-yl)-pyrimido[5,4-d]pyrimidine-2,8-diamine, 2HCl, BIBX1382BS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19ClFN7 · 2HCl · 2H2O
CAS Number:
Molecular Weight:
496.79
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

water: 1 mg/mL
DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1c(cc(cc1)Nc2ncnc3c2nc(nc3)NC4CCN(CC4)C)Cl

InChI

1S/C18H19ClFN7/c1-27-6-4-11(5-7-27)25-18-21-9-15-16(26-18)17(23-10-22-15)24-12-2-3-14(20)13(19)8-12/h2-3,8-11H,4-7H2,1H3,(H,21,25,26)(H,22,23,24)

InChI key

FTFRZXFNZVCRSK-UHFFFAOYSA-N

General description

A cell-permeable pyrimidopyrimidine compound that acts as a potent, reversible, ATP-competitive, and highly selective inhibitor of EGFR (ErbB-1, HER-1) both in cell-free enzymatic reactions (IC50 = 3 nM) and in culture (IC50 = 0.15, 1.82, and 3.2 µM in EGF-, HGF-, and FCS-dependent thymidine incorporation, respectively, in KB cells). It exhibits 1,000-fold greater selectivity over ErbB-2 (HER-2, neu; IC50 = 3.4 µM) and shows little activity towards IGF1R, β-InsRK, HGFR, c-src, and VEGFR-2 even at concentrations as high as 10 µM. Its antitumor efficacy has also been demonstrated in a murine xenograft model in vivo (daily p.o. dose 10 mg/kg).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
EGFR (ErbB-1, HER-1)
Product competes with ATP.
Reversible: yes
Target IC50: 3 nM against EGFR (ErbB-1, HER-1) in cell-free enzymatic reactions; 0.15, 1.82, and 3.2 µM in EGF-, HGF-, and FCS-dependent thymidine incorporation, respectively, in KB cells

Warning

Toxicity: Irritant (B)

Other Notes

Solca, F.F., et al. 2004. J. Pharm. Exp. Ther.311, 502.
Egeblad, M., et al. 2001. Biochem. Biophys. Res. Commun.281, 25.
Nuijen, B., et al. 2000. Int. J. Pharm.194, 261.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service