1.17357
MAS-100 Atmos® Sampling Head
for use with MAS-100 Atmos®, no. of holes, 300 autoclavable, hole diam. 0.6 mm
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41103403
NACRES:
NA.74
Recommended Products
material
anodized aluminum head
Agency
ISO 8573-7:2003
feature
no. of holes 300 autoclavable
manufacturer/tradename
MAS-100 Atmos®
hole diam.
0.6 mm
application(s)
compressed gas monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with MAS-100 Atmos®
storage temp.
no temp limit
General description
The MAS-100 Atmos® Sampling Head is designed as an accessory for MAS-100 Atmos® Compressed Gas Sampler made from anodized aluminum. It is supplied with a perforated lid for uniform impaction and convenient agar plate usage.
Each perforated lid is provided with a black marking for individual identification and traceability. The type of the head is indicated by a 3-letter code followed by a 6-digit serial number. For the 300 x 0.6 mm perforated lid for MAS-100 Atmos® the type of code is ALA. The same information is included in the 2D bar code.
Furthermore, for easy visual identification the sieve plate is colored in blue. Additional marking indicates the correct positioning for opening and closing the lid.
The default flow rate is 100 liter/min (LPM) with an impaction speed of 20 m/s attains a nominal d50 value of 1.1 μm.
An optional perforated lid with 300x0.47mm diameter is available to achieve a d50 of 1.1 μm as well at a pre-selected flow rate of 50 LPM (refer related product below)
Each perforated lid is provided with a black marking for individual identification and traceability. The type of the head is indicated by a 3-letter code followed by a 6-digit serial number. For the 300 x 0.6 mm perforated lid for MAS-100 Atmos® the type of code is ALA. The same information is included in the 2D bar code.
Furthermore, for easy visual identification the sieve plate is colored in blue. Additional marking indicates the correct positioning for opening and closing the lid.
The default flow rate is 100 liter/min (LPM) with an impaction speed of 20 m/s attains a nominal d50 value of 1.1 μm.
An optional perforated lid with 300x0.47mm diameter is available to achieve a d50 of 1.1 μm as well at a pre-selected flow rate of 50 LPM (refer related product below)
Application
The MAS-100 Atmos® Sampling head supplied with 300 x 0.6 mm holes perforated lid is used for compressed gas sampling.
Features and Benefits
- traceability by individual black marking incl. 2D bar code
- accuracy of holes allow exchange on the instrument without need for additional calibration.
- compatible with a wide range of disinfectants and autoclavable
Legal Information
MAS-100 Atmos is a registered trademark of MBV AG
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service