Skip to Content
Merck
All Photos(1)

Key Documents

1.00860

Supelco

Molybdenum Test, photometric

photometric, 0.02-1.00 mg/L (Mo), 0.03-1.67 mg/L (MoO42-), 0.04-2.15 mg/L (Na2MoO4), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Molybdenum Cell Test, photometric, 0.02-1.00 mg/L (Mo), 0.03-1.67 mg/L (MoO42-), 0.04-2.15 mg/L (Na2MoO4), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

molybdenum

measuring range

0.02-1.00 mg/L (Mo)
0.03-1.67 mg/L (MoO42-)
0.04-2.15 mg/L (Na2MoO4)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric

storage temp.

15-25°C

General description

Method: photometric 0.02 - 1.00 mg/l Mo 0.03 - 1.67 mg/l MoO₄²⁻ 0.04 - 2.15 mg/l Na₂MoO₄ Spectroquant®

This Spectroquant® Molybdenum Cell Test allows the accurate quantification of the molybdenum content in aqueous samples.

Method applied: In the presence of a cationic surfactant molybdenum(VI) reacts with bromopyrogallol red to form a blue complex which is determined photometrically.


The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.


All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service