Skip to Content
Merck
All Photos(1)

Key Documents

T-158

Supelco

(-)-11-nor-9-Carboxy-Delta8-THC-D3 solution

100 μg/mL (Degassed Methanol), certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

(6aR,10aR)-6a,7,10,10a-Tetrahydro-1-hydroxy-6,6-dimethyl-3-(pentyl-5,5,5-d3)-6H-dibenzo[b,d]pyran-9-carboxylic acid (ACI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25D3O4
CAS Number:
Molecular Weight:
347.46
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (Degassed Methanol)

format

single component solution

storage temp.

-10 to -25°C

General description

This internal standard is applicable for numerous applications in GC/MS or LC/MS testing from forensic analysis and clinical toxicology to urine drug testing and isotope dilution methods. 11-nor-9-Carboxy-Delta8-THC, is the major metabolite of Delta8-THC found in urine or blood of cannabis users.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service