Skip to Content
Merck
All Photos(2)

Key Documents

857510O

Avanti

Anteiso 15:0 fatty acid

>99% (TLC)

Synonym(s):

Anteiso 15:0 Fatty Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H30O2
CAS Number:
Molecular Weight:
242.40
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

oil

packaging

package of 1 × 10 mg
package of 1 × 25 mg

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bioactive lipids

shipped in

dry ice

storage temp.

-65 to -96°C

SMILES string

CC[C@H](C)CCCCCCCCCCC(O)=O

InChI

1S/C15H30O2/c1-3-14(2)12-10-8-6-4-5-7-9-11-13-15(16)17/h14H,3-13H2,1-2H3,(H,16,17)/t14-/m0/s1

InChI key

XKLJLHAPJBUBNL-AWEZNQCLSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

5 mL Amber Glass Screw Cap Vial (857510O-10MG)
5 mL Amber Glass Screw Cap Vial (857510O-25MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service