Skip to Content
Merck
All Photos(1)

Key Documents

840529C

Avanti

16:0-18:1 DG glucose

Avanti Research - A Croda Brand

Synonym(s):

1-palmitoyl-2-oleoyl-3-(ß-D-glucosyl)-sn-glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H80O10
CAS Number:
Molecular Weight:
757.09
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

chloroform solution

packaging

pkg of 1 × 5 mg

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

neutral lipids
neutral glycerides

shipped in

dry ice

storage temp.

−20°C

Packaging

pkg of 1 x 5mg; 1 mL (840529C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service