810229P
Avanti
18-NBD 18:1 Coenzyme A
Avanti Research™ - A Croda Brand
Synonym(s):
{N-[(7-nitro-2-1,3-benzoxadiazol-4-yl)-methyl]amino} oleoyl Coenzyme A (ammonium salt)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C45H79N14O20P3S
CAS Number:
Molecular Weight:
1261.18
UNSPSC Code:
12352211
NACRES:
NA.25
Recommended Products
Assay
>99% (TLC)
form
powder
packaging
package of 1 × 1 mg
manufacturer/tradename
Avanti Research™ - A Croda Brand
application(s)
lipidomics
lipid type
coenzymes
shipped in
dry ice
storage temp.
−20°C
Packaging
5 mL Amber Glass Screw Cap Vial (810229P-1MG)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service