MT30207959
Mettler Toledo Seven2Go™ DO meter S4
Standard Kit, polarographic
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115611
NACRES:
NB.61
Recommended Products
description
connector BNC/RCA
pH/ion/ORP meters
feature
Standard Kit, polarographic
manufacturer/tradename
Mettler-Toledo 30207959
dissolved oxygen measuring range
0.00-99.9 mg/L (ppm)
pressure measuring range
500-1100 (mbar)
saturation measuring range
0.0-600% (DO)
temperature measuring range
0-60 °C
battery power
4 x 1.5 V AA (or 1.3 V NiMH accumulators)
Looking for similar products? Visit Product Comparison Guide
General description
Simple and reliable, attractively designed and robust. The Seven2Go portables are engineered to provide users with fast quality data, one-handed operation, and durability, making them ideally suited for mobile applications in the laboratory, at-line, or outdoors.
Simple Menu – Intuitive Operation:
T-Pad – Comfortable, Fast Navigation:
ISM® Ready – Reduced Chances for Error:
IP67 – Weatherproof and Robust Design:
Simple Menu – Intuitive Operation:
- Seven2Go′s new intuitive menu guarantees out-of-the-box operation for anyone. Users no longer need to study lengthy operating instructions
T-Pad – Comfortable, Fast Navigation:
- Seven2Go′s new T-Pad improves the speed with which you move through menus. The smart button arrangement allows for faster navigation that provide you shorter time to actual measurement
ISM® Ready – Reduced Chances for Error:
- Upon connection of any InLab ISM (Intelligent Sensor Management) sensor, the instrument automatically recognizes the sensor and stores sensor ID, serial number, and last calibration data
IP67 – Weatherproof and Robust Design:
- Thanks to its IP67 protection and drop test resistance, the new Seven2Go portables are able to withstand harsh and demanding environments
Application
Aqueous samples, alcoholic and non-alcoholic beverages
Features and Benefits
Includes: Meter, InLab 605-ISM, electrode clip, base stand, instructions, quick guide, test certificate, declaration of conformity, batteries, bottles for calibration
Legal Information
ISM is a registered trademark of Mettler-Toledo, Inc.
Seven2Go is a trademark of Mettler-Toledo, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service