Skip to Content
Merck
All Photos(1)

Key Documents

GF40225980

Niobium-zirconium alloy, Nb99Zr1

200mm tube, 3.95mm outside diameter, 0.35mm wall thickness, 3.25mm inside diameter, stress relieved

Synonym(s):

NB037350

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 402-259-80

L

200 mm

O.D. × I.D. × wall thickness

3.95 mm × 3.25 mm × 0.35 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service