C87800
4-Cumylphenol
99%
Synonym(s):
4-(2-Phenylisopropyl)phenol
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
C6H5C(CH3)2C6H4OH
CAS Number:
Molecular Weight:
212.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
crystals
bp
335 °C (lit.)
mp
74-76 °C (lit.)
SMILES string
CC(C)(c1ccccc1)c2ccc(O)cc2
InChI
1S/C15H16O/c1-15(2,12-6-4-3-5-7-12)13-8-10-14(16)11-9-13/h3-11,16H,1-2H3
InChI key
QBDSZLJBMIMQRS-UHFFFAOYSA-N
Gene Information
rat ... Ar(24208)
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Yichang Chen et al.
PLoS genetics, 12(7), e1006223-e1006223 (2016-07-30)
Concerns about the safety of Bisphenol A, a chemical found in plastics, receipts, food packaging and more, have led to its replacement with substitutes now found in a multitude of consumer products. However, several popular BPA-free alternatives, such as Bisphenol
Xiaoxia Wang et al.
Environmental toxicology, 35(5), 628-638 (2020-01-10)
Bisphenol A (BPA) and 4-cumylphenol (4-CP), as estrogen-like chemicals, are ubiquitous in the environment media and associated with the occurrence and development of hormone-dependent tumors. However, the combinatorial effects of these two structurally similar alkylphenols are not well informed. In
Jaeseong Jeong et al.
International journal of molecular sciences, 20(5) (2019-03-13)
Molecular docking is used to analyze structural complexes of a target with its ligand for understanding the chemical and structural basis of target specificity. This method has the potential to be applied for discovering molecular initiating events (MIEs) in the
Zong-Da Du et al.
Talanta, 206, 120179-120179 (2019-09-14)
In this work, the magnetic amino-functionalized microporous organic network composites (Fe3O4@MON-NH2) were rational designed and facile synthesized for magnetic solid phase extraction (MSPE) of endocrine disrupting chemicals (EDCs), followed by their analysis with high-performance liquid chromatography. The incorporation of amino
Yohann Catel et al.
Dental materials : official publication of the Academy of Dental Materials, 37(2), 351-358 (2020-12-29)
To evaluate high refractive index methacrylates as diluents for the formulation of radiopaque esthetic bulk-fill composites. 2-(4-Cumylphenoxy)ethyl methacrylate 1, 2-(2-phenylphenoxy)ethyl methacrylate 2 and 2-[2-(2-phenylphenoxy)ethoxy]ethyl methacrylate 3 were synthesized and characterized by 1H NMR spectroscopy. The reactivity of each monomer was
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service