C32801
2-Chlorocyclopentanone
contains 0.2% calcium carbonate as stabilizer, 98%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
ClC5H7(=O)
CAS Number:
Molecular Weight:
118.56
Beilstein:
1098899
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
contains
0.2% calcium carbonate as stabilizer
refractive index
n20/D 1.474 (lit.)
bp
72-74 °C/12 mmHg (lit.)
density
1.185 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
ClC1CCCC1=O
InChI
1S/C5H7ClO/c6-4-2-1-3-5(4)7/h4H,1-3H2
InChI key
AXDZFGRFZOQVBV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
170.6 °F - closed cup
Flash Point(C)
77 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service